Assay Details
Target Gene Details
Entrez Gene ID: | 79953 |
Gene Name: | synapse differentiation inducing 1 |
Gene Aliases: |
C20orf39, DSPC2, IFITMD5, TMEM90B |
Location: |
Chr.20:24469199-24666617 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SYNDIG1 | NM_001323606.1 | NP_001310535.1 | ||
| NM_001323607.1 | NP_001310536.1 | |||
| NM_024893.2 | NP_079169.1 | |||
| XM_011529347.2 | XP_011527649.1 | |||
| XM_011529348.2 | XP_011527650.1 | |||
| XM_011529349.2 | XP_011527651.1 | |||
| XM_011529350.1 | XP_011527652.1 | |||
| XM_011529351.1 | XP_011527653.1 | |||
| XM_011529352.2 | XP_011527654.1 | |||
| XM_011529353.2 | XP_011527655.1 | |||
| XM_011529354.2 | XP_011527656.1 | |||
| XM_011529356.2 | XP_011527658.1 | |||
| XM_011529358.2 | XP_011527660.1 | |||
| XM_017028064.1 | 1 | 390 | XP_016883553.1 | |
| XM_017028065.1 | XP_016883554.1 | |||
| XM_017028066.1 | XP_016883555.1 | |||
| XM_017028067.1 | XP_016883556.1 | |||
| XM_017028069.1 | XP_016883558.1 | |||
| AK024282.1 | BAB14872.1 | |||
| BC030637.1 | AAH30637.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv9793 | Chr.20:23958976 - 24754214 on Build GRCh38 | Gain+Loss |
|
| nsv522873 | Chr.20:23923336 - 26134874 on Build GRCh38 | Gain |
|
| nsv833946 | Chr.20:24339739 - 24510066 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map