Assay Details
Target Gene Details
Entrez Gene ID: | 140685 |
Gene Name: | zinc finger and BTB domain containing 46 |
Gene Aliases: |
BTBD4, BZEL, RINZF, ZNF340, dJ583P15.7, dJ583P15.8 |
Location: |
Chr.20:63743668-63832332 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZBTB46 | XM_005260196.3 | XP_005260253.1 | ||
| XM_005260197.4 | XP_005260254.1 | |||
| XM_005260198.4 | XP_005260255.1 | |||
| XM_006723700.3 | XP_006723763.1 | |||
| XM_011528548.2 | XP_011526850.1 | |||
| XM_011528549.2 | XP_011526851.1 | |||
| XM_017027667.1 | XP_016883156.1 | |||
| BC052269.1 | AAH52269.1 | |||
| BC073800.1 | AAH73800.1 |
Target Gene Details
Entrez Gene ID: | 101928604 |
Gene Name: | ZBTB46 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.20:63808076-63823015 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZBTB46-AS1 | NR_110081.1 | |||
| BI089231.1 | ||||
| BX283586.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv953308 | Chr.20:63650148 - 63831847 on Build GRCh38 | Deletion |
|
| nsv428381 | Chr.20:63583375 - 64334167 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map