Assay Details
Target Gene Details
Entrez Gene ID: | 79953 |
Gene Name: | synapse differentiation inducing 1 |
Gene Aliases: |
C20orf39, DSPC2, IFITMD5, TMEM90B |
Location: |
Chr.20:24469199-24666617 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3645552 | Chr.20:24494271 - 25093181 on Build GRCh38 | Gain |
|
| nsv9793 | Chr.20:23958976 - 24754214 on Build GRCh38 | Gain+Loss |
|
| esv3645553 | Chr.20:24502289 - 24652104 on Build GRCh38 | Gain |
|
| nsv522873 | Chr.20:23923336 - 26134874 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map