Assay Details
Target Gene Details
Entrez Gene ID: | 56937 |
Gene Name: | prostate transmembrane protein, androgen induced 1 |
Gene Aliases: |
STAG1, TMEPAI |
Location: |
Chr.20:57648392-57711536 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PMEPA1 | NM_001255976.1 | NP_001242905.1 | ||
| NM_020182.4 | NP_064567.2 | |||
| NM_199169.2 | NP_954638.1 | |||
| NM_199170.2 | NP_954639.1 | |||
| NM_199171.2 | NP_954640.1 | |||
| AF224278.1 | AAF86322.1 | |||
| AF305616.1 | AAL16781.1 | |||
| AK296199.1 | ||||
| AY128643.1 | AAM89277.1 | |||
| BC015918.1 | AAH15918.2 | |||
| BC080635.1 | AAH80635.1 | |||
| BQ690750.1 | ||||
| BU196912.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2758802 | Chr.20:57595722 - 58113215 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map