Assay Details
Target Gene Details
Entrez Gene ID: | 1789 |
Gene Name: | DNA methyltransferase 3 beta |
Gene Aliases: |
ICF, ICF1, M.HsaIIIB |
Location: |
Chr.20:32762385-32809356 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DNMT3B | NM_001207055.1 | NP_001193984.1 | ||
| NM_001207056.1 | NP_001193985.1 | |||
| NM_006892.3 | NP_008823.1 | |||
| NM_175848.1 | NP_787044.1 | |||
| NM_175849.1 | NP_787045.1 | |||
| AB208880.1 | BAD92117.1 | |||
| AF156487.1 | AAD53062.1 | |||
| AF156488.1 | AAD53063.1 | |||
| AF331857.1 | AAL57040.1 | |||
| AK295176.1 | ||||
| AK299776.1 | ||||
| AK299821.1 | ||||
| AK299915.1 | ||||
| AK300857.1 | ||||
| DQ321787.1 | ABC48951.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1056411 | Chr.20:32469234 - 33355029 on Build GRCh38 | Loss |
|
| nsv833956 | Chr.20:32623070 - 32814357 on Build GRCh38 | Loss |
|
| nsv833957 | Chr.20:32755316 - 32907992 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map