Assay Details
Target Gene Details
Entrez Gene ID: | 80331 |
Gene Name: | DnaJ heat shock protein family (Hsp40) member C5 |
Gene Aliases: |
CLN4, CLN4B, CSP, DNAJC5A, NCL, mir-941-2, mir-941-3, mir-941-4, mir-941-5 |
Location: |
Chr.20:63895102-63936031 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DNAJC5 | NM_025219.2 | NP_079495.1 | ||
| AK289585.1 | ||||
| BC053642.1 | AAH53642.1 | |||
| DB452738.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509784 | Chr.20:63824382 - 63983325 on Build GRCh38 | Insertion |
|
| nsv428381 | Chr.20:63583375 - 64334167 on Build GRCh38 | Gain+Loss |
|
| esv3646330 | Chr.20:63914704 - 63918693 on Build GRCh38 | Loss |
|
| nsv469773 | Chr.20:63845428 - 64039312 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map