Assay Details
Target Gene Details
Entrez Gene ID: | 50861 |
Gene Name: | stathmin 3 |
Gene Aliases: |
SCLIP |
Location: |
Chr.20:63639705-63653610 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| STMN3 | NM_001276310.1 | NP_001263239.1 | ||
| NM_015894.3 | NP_056978.2 | |||
| NR_075070.1 | ||||
| AK074860.1 | BAC11252.1 | |||
| AK094112.1 | ||||
| AK298903.1 | ||||
| AK310010.1 | ||||
| BC009381.2 | AAH09381.1 | |||
| BC025234.1 | AAH25234.1 | |||
| DA181040.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1160690 | Chr.20:63639552 - 63744287 on Build GRCh38 | Deletion |
|
| nsv428381 | Chr.20:63583375 - 64334167 on Build GRCh38 | Gain+Loss |
|
| nsv470563 | Chr.20:63486715 - 63793308 on Build GRCh38 | Loss |
|
| esv2758806 | Chr.20:63470996 - 63745014 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map