Assay Details
Target Gene Details
Entrez Gene ID: | 5152 |
Gene Name: | phosphodiesterase 9A |
Gene Aliases: |
HSPDE9A2 |
Location: |
Chr.21:42653752-42775509 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv3534 | Chr.21:42650074 - 42681450 on Build GRCh38 | Insertion |
|
| nsv522001 | Chr.21:42421796 - 42827928 on Build GRCh38 | Gain |
|
| esv2666871 | Chr.21:42615442 - 43467915 on Build GRCh38 | Deletion |
|
| nsv587655 | Chr.21:42665548 - 42680969 on Build GRCh38 | Loss |
|
| nsv953634 | Chr.21:42648991 - 42669590 on Build GRCh38 | Deletion |
|
| esv3647086 | Chr.21:42641558 - 42685014 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map