Assay Details
Target Gene Details
Entrez Gene ID: | 388813 |
Gene Name: | uncharacterized protein ENSP00000383407-like |
Gene Aliases: |
- |
Location: |
Chr.21:14591392-14643107 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC388813 | NM_001256579.2 | NP_001243508.2 | ||
| BG221793.1 | ||||
| GR939211.1 |
Target Gene Details
Entrez Gene ID: | 100874190 |
Gene Name: | SAMSN1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.21:14582202-14598303 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SAMSN1-AS1 | NR_046512.2 | |||
| BF184654.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834046 | Chr.21:14531471 - 14705048 on Build GRCh38 | Gain+Loss |
|
| esv3646433 | Chr.21:14569026 - 14657077 on Build GRCh38 | Gain |
|
| esv3646434 | Chr.21:14570021 - 14827408 on Build GRCh38 | Gain |
|
| nsv834045 | Chr.21:14528357 - 14740529 on Build GRCh38 | Loss |
|

Genomic Map