Assay Details
Target Gene Details
Entrez Gene ID: | 1827 |
Gene Name: | regulator of calcineurin 1 |
Gene Aliases: |
ADAPT78, CSP1, DSC1, DSCR1, MCIP1, RCN1 |
Location: |
Chr.21:34516442-34615142 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RCAN1 | NM_001285389.2 | NP_001272318.1 | ||
| NM_004414.6 | NP_004405.3 | |||
| NM_203417.2 | NP_981962.1 | |||
| XM_005260929.1 | XP_005260986.1 | |||
| AF400429.1 | AAK92478.1 | |||
| AK092184.1 | ||||
| AL556803.3 | ||||
| AY325903.1 | AAP96743.1 | |||
| BC002864.2 | AAH02864.1 | |||
| BT007363.1 | AAP36027.1 | |||
| DA503835.1 | ||||
| U28833.2 | AAB81557.1 | |||
| U85265.2 | AAB84370.1 | |||
| U85266.2 | AAB84371.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv587428 | Chr.21:34515061 - 34543424 on Build GRCh38 | Loss |
|
| dgv7824n54 | Chr.21:34352440 - 34535115 on Build GRCh38 | Gain |
|
| dgv4426n100 | Chr.21:34339322 - 34534919 on Build GRCh38 | Gain |
|
| esv3575386 | Chr.21:34353927 - 34534570 on Build GRCh38 | Gain |
|
| esv2762109 | Chr.21:34350696 - 34531755 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map