Assay Details
Target Gene Details
Entrez Gene ID: | 100874322 |
Gene Name: | BACH1 intronic transcript 2 |
Gene Aliases: |
- |
Location: |
Chr.21:29372501-29373980 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BACH1-IT2 | NR_046564.2 | 2 | 344 | |
| AA912526.1 |

Genomic Map