Assay Details
Target Gene Details
Entrez Gene ID: | 1827 |
Gene Name: | regulator of calcineurin 1 |
Gene Aliases: |
ADAPT78, CSP1, DSC1, DSCR1, MCIP1, RCN1 |
Location: |
Chr.21:34516442-34615142 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv587428 | Chr.21:34515061 - 34543424 on Build GRCh38 | Loss |
|
| dgv7824n54 | Chr.21:34352440 - 34535115 on Build GRCh38 | Gain |
|
| dgv4426n100 | Chr.21:34339322 - 34534919 on Build GRCh38 | Gain |
|
| esv3575386 | Chr.21:34353927 - 34534570 on Build GRCh38 | Gain |
|
| esv2762109 | Chr.21:34350696 - 34531755 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map