Assay Details
Target Gene Details
Entrez Gene ID: | 10737 |
Gene Name: | RFPL3 antisense |
Gene Aliases: |
NCRNA00005, RFPL3-AS, RFPL3-AS1, RFPL3ANT, RFPL3AS |
Location: |
Chr.22:32359906-32371264 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RFPL3S | NR_001450.3 | |||
| NR_002596.2 | ||||
| NR_104232.1 | ||||
| AJ010233.1 | ||||
| HY013491.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3575476 | Chr.22:32073030 - 32383792 on Build GRCh38 | Gain |
|
| esv3893474 | Chr.22:32054375 - 32387314 on Build GRCh38 | Gain |
|
| nsv588895 | Chr.22:32178287 - 32470947 on Build GRCh38 | Gain |
|
| esv2422247 | Chr.22:32090461 - 32392271 on Build GRCh38 | Duplication |
|

Genomic Map