Assay Details
Target Gene Details
Entrez Gene ID: | 9814 |
Gene Name: | SFI1 centrin binding protein |
Gene Aliases: |
PISD, PPP1R139, hSfi1p |
Location: |
Chr.22:31496139-31618554 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv3594 | Chr.22:31504103 - 31537708 on Build GRCh38 | Insertion |
|
| nsv1059314 | Chr.22:31497627 - 31540102 on Build GRCh38 | Gain |
|
| esv2663679 | Chr.22:31499437 - 31515090 on Build GRCh38 | Deletion |
|
| nsv588894 | Chr.22:31437773 - 31581608 on Build GRCh38 | Loss |
|
| esv3647589 | Chr.22:31499443 - 31515048 on Build GRCh38 | Loss |
|
| nsv1063634 | Chr.22:31401018 - 31525268 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map