Assay Details
Target Gene Details
Entrez Gene ID: | 1727 |
Gene Name: | cytochrome b5 reductase 3 |
Gene Aliases: |
B5R, DIA1 |
Location: |
Chr.22:42617840-42649399 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CYB5R3 | NM_000398.6 | NP_000389.1 | ||
| NM_001129819.2 | NP_001123291.1 | |||
| NM_001171660.1 | NP_001165131.1 | |||
| NM_001171661.1 | NP_001165132.1 | |||
| NM_007326.4 | NP_015565.1 | |||
| AF361370.1 | AAL87744.1 | |||
| AJ010116.1 | CAA09006.1 | |||
| AJ010117.1 | CAA09007.1 | |||
| AJ010118.1 | CAA09008.1 | |||
| AJ310899.1 | CAC84523.1 | |||
| AJ310900.1 | CAC84524.1 | |||
| AK124528.1 | BAC85875.1 | |||
| AK302204.1 | ||||
| BC004821.1 | AAH04821.1 | |||
| BM564146.1 | ||||
| BT009821.1 | AAP88823.1 | |||
| CR456435.1 | CAG30321.1 | |||
| M16462.1 | AAA52307.1 | |||
| Y09501.1 | CAA70696.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv524028 | Chr.22:42592111 - 42631818 on Build GRCh38 | Loss |
|
| esv2758839 | Chr.22:42371195 - 42687300 on Build GRCh38 | Gain+Loss |
|
| nsv428391 | Chr.22:42371295 - 42698202 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map