Assay Details
Target Gene Details
Entrez Gene ID: | 100132273 |
Gene Name: | NDUFA6 antisense RNA 1 (head to head) |
Gene Aliases: |
- |
Location: |
Chr.22:42090933-42125349 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NDUFA6-AS1 | NR_034118.2 | |||
| AK123630.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv508737 | Chr.22:42052007 - 42134336 on Build GRCh38 | Insertion |
|
| nsv3642 | Chr.22:42115497 - 42128296 on Build GRCh38 | Insertion |
|
| esv2674409 | Chr.22:42117842 - 42120689 on Build GRCh38 | Deletion |
|
| nsv834210 | Chr.22:42057329 - 42209100 on Build GRCh38 | Loss |
|
| esv3647808 | Chr.22:42117846 - 42120664 on Build GRCh38 | Loss |
|
| nsv3641 | Chr.22:42099466 - 42146296 on Build GRCh38 | Deletion |
|

Genomic Map