Assay Details
Target Gene Details
Entrez Gene ID: | 26150 |
Gene Name: | RIB43A domain with coiled-coils 2 |
Gene Aliases: |
C22orf11, TRIB |
Location: |
Chr.22:45413691-45432422 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RIBC2 | NM_015653.4 | NP_056468.3 | ||
| XM_005261524.4 | XP_005261581.1 | |||
| XM_017028766.1 | XP_016884255.1 | |||
| AK098586.1 | ||||
| AK225722.1 | ||||
| AL442116.1 | CAC09526.1 | |||
| BC003024.1 | AAH03024.1 | |||
| BC009904.2 | AAH09904.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1066204 | Chr.22:45254728 - 45870451 on Build GRCh38 | Gain |
|
| nsv834221 | Chr.22:45383909 - 45578311 on Build GRCh38 | Loss |
|
| nsv3651 | Chr.22:45403708 - 45435552 on Build GRCh38 | Insertion |
|
| esv34218 | Chr.22:45254944 - 45515710 on Build GRCh38 | Gain |
|
| esv3647931 | Chr.22:45382351 - 45517601 on Build GRCh38 | Gain |
|
| esv2758843 | Chr.22:45178343 - 45529286 on Build GRCh38 | Gain |
|
| nsv9921 | Chr.22:45247876 - 45514132 on Build GRCh38 | Gain+Loss |
|
| esv3647921 | Chr.22:45247581 - 45517162 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs114380830] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map