Assay Details
Target Gene Details
Entrez Gene ID: | 102466081 |
Gene Name: | microRNA 5739 |
Gene Aliases: |
hsa-mir-5739 |
Location: |
Chr.22:28459869-28459948 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MIR5739 | NR_106713.1 |
Target Gene Details
Entrez Gene ID: | 23331 |
Gene Name: | tetratricopeptide repeat domain 28 |
Gene Aliases: |
TPRBK |
Location: |
Chr.22:27978014-28679865 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TTC28 | NM_001145418.1 | NP_001138890.1 | ||
| XM_005261405.2 | XP_005261462.1 | |||
| XM_011530018.2 | XP_011528320.1 | |||
| XM_011530019.2 | XP_011528321.1 | |||
| XM_011530020.1 | XP_011528322.1 | |||
| XM_011530021.2 | XP_011528323.1 | |||
| XM_011530022.1 | XP_011528324.1 | |||
| XM_017028673.1 | XP_016884162.1 | |||
| AB665284.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv588880 | Chr.22:28411289 - 28605801 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map