Assay Details
Target Gene Details
Entrez Gene ID: | 25829 |
Gene Name: | transmembrane protein 184B |
Gene Aliases: |
C22orf5, FM08, HS5O6A, HSPC256 |
Location: |
Chr.22:38216045-38273034 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 12 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834194 | Chr.22:38063495 - 38240504 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map