Assay Details
Target Gene Details
Entrez Gene ID: | 7332 |
Gene Name: | ubiquitin conjugating enzyme E2 L3 |
Gene Aliases: |
E2-F1, L-UBC, UBCH7, UbcM4 |
Location: |
Chr.22:21549447-21624034 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UBE2L3 | NM_001256355.1 | NP_001243284.1 | ||
| NM_001256356.1 | NP_001243285.1 | |||
| NM_003347.3 | NP_003338.1 | |||
| NR_028436.2 | ||||
| NR_046082.1 | ||||
| XM_017028935.1 | XP_016884424.1 | |||
| AJ000519.1 | CAA04156.1 | |||
| AK125396.1 | ||||
| AK291083.1 | ||||
| AK293179.1 | ||||
| AK299985.1 | ||||
| AK311761.1 | ||||
| BC053368.1 | AAH53368.1 | |||
| BU567707.1 | ||||
| BU584304.1 | ||||
| CD690076.1 | ||||
| CR456606.1 | CAG30492.1 | |||
| DA271653.1 | ||||
| S81003.1 | AAB36017.1 | |||
| X92962.1 | CAA63538.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv428386 | Chr.22:21328921 - 21595800 on Build GRCh38 | Loss |
|
| esv3647318 | Chr.22:21567924 - 21611833 on Build GRCh38 | Gain |
|
| nsv834145 | Chr.22:21420780 - 21595800 on Build GRCh38 | Loss |
|
| nsv1160730 | Chr.22:21373849 - 21633730 on Build GRCh38 | Deletion |
|
| dgv4491n100 | Chr.22:21454896 - 21597273 on Build GRCh38 | Gain |
|
| dgv4488n100 | Chr.22:21212930 - 21597273 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map