Assay Details
Target Gene Details
Entrez Gene ID: | 101927447 |
Gene Name: | uncharacterized LOC101927447 |
Gene Aliases: |
- |
Location: |
Chr.22:43275942-43283830 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101927447 | NR_134582.1 | |||
| DA931830.1 |
Target Gene Details
Entrez Gene ID: | 80274 |
Gene Name: | signal peptide, CUB domain and EGF like domain containing 1 |
Gene Aliases: |
- |
Location: |
Chr.22:43203223-43343388 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SCUBE1 | NM_173050.3 | NP_766638.2 | ||
| XM_005261752.1 | XP_005261809.1 | |||
| XM_011530389.2 | XP_011528691.1 | |||
| XM_011530390.2 | XP_011528692.1 | |||
| AF525689.1 | AAN77133.1 | |||
| AK127719.1 | BAC87098.1 | |||
| BC047916.1 | AAH47916.1 | |||
| BC127263.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv955166 | Chr.22:43238095 - 43320894 on Build GRCh38 | Deletion |
|
| nsv834216 | Chr.22:43184904 - 43383781 on Build GRCh38 | Loss |
|
| nsv520927 | Chr.22:43154143 - 43307985 on Build GRCh38 | Loss |
|
| nsv589089 | Chr.22:43274380 - 43338260 on Build GRCh38 | Gain |
|
| esv33816 | Chr.22:43113758 - 43326122 on Build GRCh38 | Gain |
|
| dgv844e214 | Chr.22:43185465 - 43295588 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs74364212] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map