Assay Details
Target Gene Details
Entrez Gene ID: | 388906 |
Gene Name: | opioid growth factor receptor pseudogene 1 |
Gene Aliases: |
- |
Location: |
Chr.22:42269753-42274862 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OGFRP1 | NR_036498.1 | 2 | 567 | |
| BC039496.1 | 2 | 567 | AAH39496.1 |
Target Gene Details
Entrez Gene ID: | 6942 |
Gene Name: | transcription factor 20 |
Gene Aliases: |
AR1, SPBP, TCF-20 |
Location: |
Chr.22:42160013-42283927 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TCF20 | NM_005650.3 | NP_005641.1 | ||
| KF851355.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829244 | Chr.22:42253613 - 42380506 on Build GRCh38 | Gain |
|
| nsv834211 | Chr.22:42209111 - 42386001 on Build GRCh38 | Loss |
|
| nsv955165 | Chr.22:42270495 - 42344594 on Build GRCh38 | Deletion |
|
| esv3647816 | Chr.22:42271998 - 42279309 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map