Assay Details
Target Gene Details
Entrez Gene ID: | 730005 |
Gene Name: | SEC14 like lipid binding 6 |
Gene Aliases: |
- |
Location: |
Chr.22:30522797-30546717 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SEC14L6 | NM_001193336.2 | NP_001180265.2 | ||
| XM_011530357.2 | 13 | 1631 | XP_011528659.1 | |
| XM_011530358.2 | 12 | 1535 | XP_011528660.1 | |
| XM_011530360.2 | 12 | 1442 | XP_011528662.1 | |
| XM_017028931.1 | 13 | 1388 | XP_016884420.1 | |
| XM_017028932.1 | 12 | 1507 | XP_016884421.1 | |
| XM_017028933.1 | 11 | 1122 | XP_016884422.1 | |
| XM_017028934.1 | 11 | 1336 | XP_016884423.1 | |
| BX283471.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829171 | Chr.22:30202041 - 30647591 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map