Assay Details
Target Gene Details
Entrez Gene ID: | 101928578 |
Gene Name: | RAP2C antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.X:132218507-132432811 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RAP2C-AS1 | NR_110410.1 | |||
| AK095439.1 | ||||
| CA867598.1 |

Genomic Map