Assay Details
Target Gene Details
Entrez Gene ID: | 55796 |
Gene Name: | muscleblind like splicing regulator 3 |
Gene Aliases: |
CHCR, MBLX, MBLX39, MBXL |
Location: |
Chr.X:132369314-132490030 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MBNL3 | NM_001170701.1 | 8 | 7844 | NP_001164172.1 |
| NM_001170702.1 | 7 | 7808 | NP_001164173.1 | |
| NM_001170703.1 | 8 | 7777 | NP_001164174.1 | |
| NM_001170704.1 | 9 | 8044 | NP_001164175.1 | |
| NM_018388.3 | 8 | 8007 | NP_060858.2 | |
| NM_133486.2 | 9 | 7976 | NP_597846.1 | |
| XM_005262431.3 | 10 | 8893 | XP_005262488.1 | |
| XM_005262434.3 | 9 | 8814 | XP_005262491.1 | |
| XM_005262436.3 | 9 | 8781 | XP_005262493.1 | |
| XM_005262437.3 | 9 | 7919 | XP_005262494.1 | |
| XM_005262438.3 | 8 | 7883 | XP_005262495.1 | |
| XM_005262439.3 | 9 | 7851 | XP_005262496.1 | |
| XM_005262440.3 | 11 | 8938 | XP_005262497.1 | |
| XM_005262441.3 | 9 | 8042 | XP_005262498.1 | |
| XM_005262442.3 | 9 | 8013 | XP_005262499.1 | |
| XM_011531364.2 | 8 | 8929 | XP_011529666.1 | |
| XM_017029632.1 | 10 | 16323 | XP_016885121.1 | |
| XM_017029633.1 | 11 | 9058 | XP_016885122.1 | |
| XM_017029634.1 | 9 | 16287 | XP_016885123.1 | |
| XM_017029635.1 | 9 | 16249 | XP_016885124.1 | |
| XM_017029636.1 | 9 | 8748 | XP_016885125.1 | |
| XM_017029637.1 | 8 | 16213 | XP_016885126.1 | |
| XM_017029638.1 | 11 | 8956 | XP_016885127.1 | |
| CR627122.1 | 1 | 4389 |
Target Gene Details
Entrez Gene ID: | 101928578 |
Gene Name: | RAP2C antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.X:132218507-132432811 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RAP2C-AS1 | NR_110410.1 | |||
| AK095439.1 | ||||
| CA867598.1 |
More Information
Set Membership: |
|

Genomic Map