Assay Details
Target Gene Details
Entrez Gene ID: | 55796 |
Gene Name: | muscleblind like splicing regulator 3 |
Gene Aliases: |
CHCR, MBLX, MBLX39, MBXL |
Location: |
Chr.X:132369314-132490030 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MBNL3 | NM_001170701.1 | 8 | 3405 | NP_001164172.1 |
| NM_001170702.1 | 7 | 3369 | NP_001164173.1 | |
| NM_001170703.1 | 8 | 3338 | NP_001164174.1 | |
| NM_001170704.1 | 9 | 3605 | NP_001164175.1 | |
| NM_018388.3 | 8 | 3568 | NP_060858.2 | |
| NM_133486.2 | 9 | 3537 | NP_597846.1 | |
| XM_005262431.3 | 10 | 4454 | XP_005262488.1 | |
| XM_005262434.3 | 9 | 4375 | XP_005262491.1 | |
| XM_005262436.3 | 9 | 4342 | XP_005262493.1 | |
| XM_005262437.3 | 9 | 3480 | XP_005262494.1 | |
| XM_005262438.3 | 8 | 3444 | XP_005262495.1 | |
| XM_005262439.3 | 9 | 3412 | XP_005262496.1 | |
| XM_005262440.3 | 11 | 4499 | XP_005262497.1 | |
| XM_005262441.3 | 9 | 3603 | XP_005262498.1 | |
| XM_005262442.3 | 9 | 3574 | XP_005262499.1 | |
| XM_011531364.2 | 8 | 4490 | XP_011529666.1 | |
| XM_017029632.1 | 10 | 11884 | XP_016885121.1 | |
| XM_017029633.1 | 11 | 4619 | XP_016885122.1 | |
| XM_017029634.1 | 9 | 11848 | XP_016885123.1 | |
| XM_017029635.1 | 9 | 11810 | XP_016885124.1 | |
| XM_017029636.1 | 9 | 4309 | XP_016885125.1 | |
| XM_017029637.1 | 8 | 11774 | XP_016885126.1 | |
| XM_017029638.1 | 11 | 4517 | XP_016885127.1 | |
| CR627122.1 |
Target Gene Details
Entrez Gene ID: | 101928578 |
Gene Name: | RAP2C antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.X:132218507-132432811 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RAP2C-AS1 | NR_110410.1 | |||
| AK095439.1 | ||||
| CA867598.1 |
More Information
Set Membership: |
|

Genomic Map