Assay Details
Target Gene Details
Entrez Gene ID: | 54328 |
Gene Name: | G protein-coupled receptor 173 |
Gene Aliases: |
SREB3 |
Location: |
Chr.X:53049324-53080615 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPR173 | NM_018969.5 | NP_061842.1 | ||
| XM_011530798.2 | XP_011529100.1 | |||
| BC009861.2 | AAH09861.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2740175 | Chr.X:52862926 - 55652533 on Build GRCh38 | Deletion |
|
| nsv508773 | Chr.X:53034408 - 53141410 on Build GRCh38 | Insertion |
|
| nsv435891 | Chr.X:52861979 - 55653313 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map