Assay Details
Target Gene Details
Entrez Gene ID: | 2623 |
Gene Name: | GATA binding protein 1 |
Gene Aliases: |
ERYF1, GATA-1, GF-1, GF1, NF-E1, NFE1, XLANP, XLTDA, XLTT |
Location: |
Chr.X:48786540-48794311 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GATA1 | NM_002049.3 | NP_002040.1 | ||
| XM_011543897.2 | XP_011542199.1 | |||
| XM_011543898.2 | XP_011542200.1 | |||
| BC009797.1 | AAH09797.1 | |||
| M30601.1 | AAA35885.1 | |||
| X17254.1 | CAA35120.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv528292 | Chr.X:48433286 - 49274368 on Build GRCh38 | Gain |
|
| esv32909 | Chr.X:48003374 - 49285646 on Build GRCh38 | Gain+Loss |
|
| esv3576862 | Chr.X:48755018 - 48837713 on Build GRCh38 | Gain |
|
| nsv6893 | Chr.X:48778273 - 48810147 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map