Assay Details
Target Gene Details
Entrez Gene ID: | 10251 |
Gene Name: | sprouty RTK signaling antagonist 3 |
Gene Aliases: |
spry-3 |
Location: |
Chr.X:155612565-155782459 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SPRY3 | NM_001304990.1 | NP_001291919.1 | ||
| DA102958.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv33784 | Chr.X:153962665 - 155699588 on Build GRCh38 | Gain+Loss |
|
| nsv519485 | Chr.X:155507715 - 155699751 on Build GRCh38 | Loss |
|
| nsv520699 | Chr.X:155545002 - 155699751 on Build GRCh38 | Gain |
|
| esv2657377 | Chr.X:155641695 - 155958434 on Build GRCh38 | Deletion |
|
| esv29972 | Chr.X:154781072 - 155699751 on Build GRCh38 | Gain |
|
| esv2662461 | Chr.X:155646634 - 155651390 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map