Assay Details
Target Gene Details
Entrez Gene ID: | 65991 |
Gene Name: | FUN14 domain containing 2 |
Gene Aliases: |
DC44, HCBP6, HCC3, PD03104 |
Location: |
Chr.X:155026789-155056916 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FUNDC2 | NM_023934.3 | NP_076423.2 | ||
| AF320778.1 | AAN51931.1 | |||
| AJ272054.1 | CAC81242.1 | |||
| AK091092.1 | ||||
| AK313143.1 | ||||
| AY032594.1 | AAK51136.1 | |||
| BC000255.1 | AAH00255.2 | |||
| BC072685.1 | AAH72685.1 | |||
| BC108657.1 | AAI08658.1 | |||
| BI463291.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv33784 | Chr.X:153962665 - 155699588 on Build GRCh38 | Gain+Loss |
|
| esv29972 | Chr.X:154781072 - 155699751 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map