Assay Details
Target Gene Details
Entrez Gene ID: | 55796 |
Gene Name: | muscleblind like splicing regulator 3 |
Gene Aliases: |
CHCR, MBLX, MBLX39, MBXL |
Location: |
Chr.X:132369314-132490030 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MBNL3 | NM_001170701.1 | 8 | 10315 | NP_001164172.1 |
| NM_001170702.1 | 7 | 10279 | NP_001164173.1 | |
| NM_001170703.1 | 8 | 10248 | NP_001164174.1 | |
| NM_001170704.1 | 9 | 10515 | NP_001164175.1 | |
| NM_018388.3 | 8 | 10478 | NP_060858.2 | |
| NM_133486.2 | 9 | 10447 | NP_597846.1 |
Target Gene Details
Entrez Gene ID: | 101928578 |
Gene Name: | RAP2C antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.X:132218507-132432811 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RAP2C-AS1 | NR_110410.1 | |||
| AK095439.1 | ||||
| CA867598.1 |
More Information
Set Membership: |
|

Genomic Map