Assay Details
Target Gene Details
Entrez Gene ID: | 4931 |
Gene Name: | nuclear VCP-like |
Gene Aliases: |
NVL2 |
Location: |
Chr.1:224227334-224330201 on Build GRCh38 |
Assay Gene Location: | Within Intron 27 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NVL | NM_001243146.1 | NP_001230075.1 | ||
| NM_001243147.1 | NP_001230076.1 | |||
| NM_002533.3 | NP_002524.2 | |||
| NM_206840.2 | NP_996671.1 | |||
| XM_011544196.1 | XP_011542498.1 | |||
| XM_011544198.1 | XP_011542500.1 | |||
| XM_011544199.1 | XP_011542501.1 | |||
| XM_011544200.1 | XP_011542502.1 | |||
| XM_017001378.1 | XP_016856867.1 | |||
| XM_017001379.1 | XP_016856868.1 | |||
| XM_017001381.1 | XP_016856870.1 | |||
| XM_017001382.1 | XP_016856871.1 | |||
| XM_017001383.1 | XP_016856872.1 | |||
| XM_017001386.1 | XP_016856875.1 | |||
| AK297070.1 | ||||
| AK297396.1 | ||||
| AK298244.1 | ||||
| BC012105.1 | AAH12105.1 | |||
| BU683548.1 | ||||
| U68140.1 | AAB70457.1 | |||
| U78772.1 | AAB70460.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv8868 | Chr.1:224195321 - 224564829 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map