Assay Details
Target Gene Details
Entrez Gene ID: | 8515 |
Gene Name: | integrin subunit alpha 10 |
Gene Aliases: |
PRO827 |
Location: |
Chr.1:145891208-145910189 on Build GRCh38 |
Assay Gene Location: | Within Intron 26 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ITGA10 | NM_001303040.1 | NP_001289969.1 | ||
| NM_001303041.1 | NP_001289970.1 | |||
| NM_003637.4 | NP_003628.2 | |||
| XM_005277436.3 | XP_005277493.1 | |||
| XM_011510083.2 | XP_011508385.1 | |||
| XM_011510084.1 | XP_011508386.1 | |||
| XM_017002622.1 | XP_016858111.1 | |||
| XM_017002623.1 | XP_016858112.1 | |||
| XM_017002624.1 | XP_016858113.1 | |||
| XM_017002625.1 | XP_016858114.1 | |||
| XM_017002626.1 | XP_016858115.1 | |||
| XM_017002628.1 | XP_016858117.1 | |||
| AF074015.1 | AAC31952.1 | |||
| AF112345.1 | AAF21944.1 | |||
| AK304155.1 | ||||
| AK304183.1 | ||||
| AK314255.1 | ||||
| BC140831.1 | ||||
| BC144637.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1005055 | Chr.1:145642988 - 148991068 on Build GRCh38 | Loss |
|
| nsv831381 | Chr.1:145784568 - 145960930 on Build GRCh38 | Gain |
|
| esv3587420 | Chr.1:145824369 - 145905171 on Build GRCh38 | Gain |
|
| nsv2710 | Chr.1:145894631 - 145927475 on Build GRCh38 | Insertion |
|
| esv3587419 | Chr.1:145893340 - 145919968 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map