Assay Details
Target Gene Details
Entrez Gene ID: | 163486 |
Gene Name: | DENN domain containing 1B |
Gene Aliases: |
C1ORF18, C1orf218, FAM31B |
Location: |
Chr.1:197504748-197782138 on Build GRCh38 |
Assay Gene Location: | Within Exon 30 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DENND1B | NM_001195215.1 | 23 | 6450 | NP_001182144.1 |
| NR_125340.1 | 20 | 6197 | ||
| XM_005244931.2 | 22 | 6390 | XP_005244988.1 | |
| XM_006711192.2 | 24 | 6360 | XP_006711255.1 | |
| XM_006711193.3 | 24 | 6492 | XP_006711256.1 | |
| XM_006711194.3 | 11 | 16488 | XP_006711257.1 | |
| XM_011509246.2 | 23 | 6311 | XP_011507548.1 | |
| XM_011509247.1 | 22 | 6251 | XP_011507549.1 | |
| XM_011509248.2 | 23 | 6244 | XP_011507550.1 | |
| XM_011509249.2 | 24 | 6549 | XP_011507551.1 | |
| XM_017000470.1 | 24 | 6438 | XP_016855959.1 | |
| XM_017000471.1 | 23 | 6300 | XP_016855960.1 | |
| XM_017000472.1 | 19 | 5924 | XP_016855961.1 | |
| AK093982.1 | 1 | 343 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv4010 | Chr.1:197502333 - 197542267 on Build GRCh38 | Deletion |
|
| nsv952633 | Chr.1:197503471 - 197513670 on Build GRCh38 | Duplication |
|
| nsv521647 | Chr.1:197495318 - 197560088 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map