Assay Details
Target Gene Details
Entrez Gene ID: | 5142 |
Gene Name: | phosphodiesterase 4B |
Gene Aliases: |
DPDE4, PDEIVB |
Location: |
Chr.1:65792510-66374579 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PDE4B | NM_001037341.1 | NP_001032418.1 | ||
| NM_001297440.1 | NP_001284369.1 | |||
| NM_002600.3 | NP_002591.2 | |||
| AK289969.1 | ||||
| BC101480.1 | AAI01481.1 | |||
| BC105040.1 | AAI05041.1 | |||
| BF513098.1 | ||||
| CR749667.1 | CAH18458.2 | |||
| DA103627.1 | ||||
| DA126733.1 | ||||
| L20966.1 | AAA03589.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2584680 | Chr.1:65797548 - 65799180 on Build GRCh38 | Deletion |
|
| dgv3n17 | Chr.1:65756073 - 65806453 on Build GRCh38 | Loss |
|
| nsv437216 | Chr.1:65791733 - 65806453 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map