Assay Details
Target Gene Details
Entrez Gene ID: | 79699 |
Gene Name: | zyg-11 family member B, cell cycle regulator |
Gene Aliases: |
ZYG11 |
Location: |
Chr.1:52726454-52827342 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZYG11B | NM_024646.2 | NP_078922.1 | ||
| XM_006710898.3 | XP_006710961.1 | |||
| XM_017002336.1 | XP_016857825.1 | |||
| AB051517.1 | ||||
| AK095643.1 | ||||
| AK296460.1 | ||||
| DQ646393.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1001535 | Chr.1:52620368 - 53346539 on Build GRCh38 | Gain |
|
| esv2758934 | Chr.1:52660343 - 52823509 on Build GRCh38 | Loss |
|
| nsv829859 | Chr.1:52723470 - 52925273 on Build GRCh38 | Loss |
|
| nsv1000097 | Chr.1:52743770 - 52809024 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map