Assay Details
Target Gene Details
Entrez Gene ID: | 647135 |
Gene Name: | SLIT-ROBO Rho GTPase activating protein 2B |
Gene Aliases: |
SRGAP2L, SRGAP2P2 |
Location: |
Chr.1:144887191-145093441 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SRGAP2B | NM_001271870.2 | NP_001258799.2 | ||
| XM_005277421.3 | XP_005277478.1 | |||
| XM_017002085.1 | XP_016857574.1 | |||
| XM_017002086.1 | XP_016857575.1 | |||
| XM_017002087.1 | XP_016857576.1 | |||
| XM_017002088.1 | XP_016857577.1 | |||
| XM_017002089.1 | XP_016857578.1 | |||
| XM_017002090.1 | XP_016857579.1 | |||
| XM_017002091.1 | XP_016857580.1 | |||
| XM_017002092.1 | XP_016857581.1 | |||
| BY795901.2 | ||||
| HY154277.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1136032 | Chr.1:144931440 - 144932997 on Build GRCh38 | Deletion |
|
| esv2762186 | Chr.1:144928063 - 145023143 on Build GRCh38 | Loss |
|
| nsv7947 | Chr.1:144915703 - 145001208 on Build GRCh38 | Loss |
|
| nsv946222 | Chr.1:144914153 - 145023143 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map