Assay Details
Target Gene Details
Entrez Gene ID: | 116372 |
Gene Name: | LY6/PLAUR domain containing 1 |
Gene Aliases: |
LYPDC1, PHTS |
Location: |
Chr.2:132643280-132671579 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LYPD1 | NM_001077427.3 | NP_001070895.1 | ||
| NM_001321234.1 | NP_001308163.1 | |||
| NM_001321235.1 | NP_001308164.1 | |||
| NM_144586.6 | NP_653187.3 | |||
| XM_011510546.2 | XP_011508848.1 | |||
| AK075487.1 | BAC11647.1 | |||
| AK094501.1 | BAC04368.1 | |||
| AK123029.1 | BAC85518.1 | |||
| AK130005.1 | BAC85273.1 | |||
| AK309668.1 | ||||
| AK309849.1 | ||||
| AY358628.1 | AAQ88991.1 | |||
| BC017318.2 | AAH17318.1 | |||
| BE386060.1 | ||||
| BP276862.1 | ||||
| CB110132.1 | ||||
| DA776269.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv583158 | Chr.2:132558519 - 133104527 on Build GRCh38 | Gain |
|
| nsv583152 | Chr.2:132369502 - 133631550 on Build GRCh38 | Gain |
|
| nsv1011421 | Chr.2:132137668 - 132924612 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs75763384] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map