Assay Details
Target Gene Details
Entrez Gene ID: | 130507 |
Gene Name: | ubiquitin protein ligase E3 component n-recognin 3 (putative) |
Gene Aliases: |
ZNF650 |
Location: |
Chr.2:169827463-170084131 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UBR3 | NM_172070.3 | NP_742067.3 | ||
| XM_005246299.4 | XP_005246356.1 | |||
| XM_005246300.4 | XP_005246357.1 | |||
| XM_006712268.3 | XP_006712331.1 | |||
| XM_006712269.3 | XP_006712332.1 | |||
| XM_006712270.3 | XP_006712333.2 | |||
| XM_006712271.3 | XP_006712334.1 | |||
| XM_011510604.2 | XP_011508906.1 | |||
| XM_017003356.1 | XP_016858845.1 | |||
| XM_017003357.1 | XP_016858846.1 | |||
| XM_017003358.1 | XP_016858847.1 | |||
| XM_017003359.1 | XP_016858848.1 | |||
| AI142128.1 | ||||
| BF446017.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3593331 | Chr.2:169847386 - 169853401 on Build GRCh38 | Loss |
|
| nsv583608 | Chr.2:169769815 - 169984892 on Build GRCh38 | Gain |
|
| nsv10201 | Chr.2:169701593 - 170169904 on Build GRCh38 | Loss |
|
| nsv583607 | Chr.2:169727686 - 169868204 on Build GRCh38 | Gain |
|
| nsv1011368 | Chr.2:169423740 - 170042296 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map