Assay Details
Target Gene Details
Entrez Gene ID: | 28999 |
Gene Name: | Kruppel like factor 15 |
Gene Aliases: |
KKLF |
Location: |
Chr.3:126288121-126357393 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KLF15 |

Genomic Map