Assay Details
Target Gene Details
Entrez Gene ID: | 2257 |
Gene Name: | fibroblast growth factor 12 |
Gene Aliases: |
FGF12B, FHF1 |
Location: |
Chr.3:192139393-192727599 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FGF12 | NM_004113.5 | NP_004104.3 | ||
| NM_021032.4 | NP_066360.1 | |||
| XM_005247227.2 | XP_005247284.1 | |||
| XM_006713538.3 | XP_006713601.1 | |||
| XM_017005879.1 | XP_016861368.1 | |||
| AK125307.1 | ||||
| AK312513.1 | ||||
| AK313671.1 | ||||
| BC022524.1 | AAH22524.1 | |||
| DA521146.1 | ||||
| U66197.1 | AAB18913.1 | |||
| U76381.2 | AAB18786.3 | |||
| Z70276.1 | CAA94240.1 |
Target Gene Details
Entrez Gene ID: | 100873986 |
Gene Name: | FGF12 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.3:192238630-192283097 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FGF12-AS1 | NR_046596.1 | |||
| DB034608.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2726410 | Chr.3:192219587 - 192270877 on Build GRCh38 | Deletion |
|
| dgv966e214 | Chr.3:192218451 - 192270994 on Build GRCh38 | Loss |
|
| nsv435755 | Chr.3:192218493 - 192272921 on Build GRCh38 | Deletion |
|
| nsv4170 | Chr.3:192180843 - 192272191 on Build GRCh38 | Deletion |
|
| esv3569197 | Chr.3:192259290 - 192298866 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map