Assay Details
Target Gene Details
Entrez Gene ID: | 114112 |
Gene Name: | thioredoxin reductase 3 |
Gene Aliases: |
TGR, TR2, TRXR3 |
Location: |
Chr.3:126607052-126655124 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TXNRD3 | NM_001173513.1 | NP_001166984.1 | ||
| NM_052883.1 | NP_443115.1 | |||
| AF133519.1 | AAD39929.1 | |||
| AF171055.1 | AAD51325.1 | |||
| AK299502.1 | ||||
| BC030028.2 | AAH30028.1 | |||
| BC050032.1 | AAH50032.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv7947 | Chr.3:126639812 - 126640679 on Build GRCh38 | Loss |
|
| esv2585777 | Chr.3:126639341 - 126640884 on Build GRCh38 | Deletion |
|
| esv2005941 | Chr.3:126639892 - 126640615 on Build GRCh38 | Deletion |
|
| esv987942 | Chr.3:126640016 - 126640981 on Build GRCh38 | Deletion |
|
| dgv4852n100 | Chr.3:126607261 - 126669480 on Build GRCh38 | Gain |
|
| nsv460842 | Chr.3:126614957 - 126669032 on Build GRCh38 | Gain |
|
| esv2759 | Chr.3:126640024 - 126640592 on Build GRCh38 | Loss |
|
| nsv829718 | Chr.3:126629911 - 126809810 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs139572502,rs76097983] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map