Assay Details
Target Gene Details
Entrez Gene ID: | 348808 |
Gene Name: | NPHP3 antisense RNA 1 |
Gene Aliases: |
NCRNA00119 |
Location: |
Chr.3:132722342-132874206 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NPHP3-AS1 | NR_002811.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv591798 | Chr.3:132626804 - 132975557 on Build GRCh38 | Loss |
|
| nsv591799 | Chr.3:132662755 - 133080365 on Build GRCh38 | Gain |
|
| esv3893774 | Chr.3:132705936 - 132933476 on Build GRCh38 | Loss |
|
| nsv998211 | Chr.3:132605119 - 132988974 on Build GRCh38 | Loss |
|

Genomic Map