Assay Details
Target Gene Details
Entrez Gene ID: | 101927374 |
Gene Name: | uncharacterized LOC101927374 |
Gene Aliases: |
- |
Location: |
Chr.3:79411727-79435227 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101927374 | NR_110133.1 | |||
| AK094694.1 |
Target Gene Details
Entrez Gene ID: | 6091 |
Gene Name: | roundabout guidance receptor 1 |
Gene Aliases: |
DUTT1, SAX3 |
Location: |
Chr.3:78597238-79767998 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ROBO1 | NM_002941.3 | NP_002932.1 | ||
| XM_011533976.1 | XP_011532278.1 | |||
| XM_011533977.2 | XP_011532279.1 | |||
| XM_011533978.1 | XP_011532280.1 | |||
| XM_011533979.1 | XP_011532281.1 | |||
| XM_011533980.1 | XP_011532282.1 | |||
| XM_017006982.1 | XP_016862471.1 | |||
| XM_017006984.1 | XP_016862473.1 | |||
| AF040990.1 | AAC39575.1 | |||
| AK297481.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834747 | Chr.3:79392061 - 79542773 on Build GRCh38 | Gain |
|
| esv2763281 | Chr.3:79197211 - 79440615 on Build GRCh38 | Loss |
|
| esv2422412 | Chr.3:79234377 - 81254205 on Build GRCh38 | Deletion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs116208984] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map