Assay Details
Target Gene Details
Entrez Gene ID: | 348808 |
Gene Name: | NPHP3 antisense RNA 1 |
Gene Aliases: |
NCRNA00119 |
Location: |
Chr.3:132722342-132874206 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NPHP3-AS1 | NR_002811.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv591798 | Chr.3:132626804 - 132975557 on Build GRCh38 | Loss |
|
| nsv591799 | Chr.3:132662755 - 133080365 on Build GRCh38 | Gain |
|
| nsv998211 | Chr.3:132605119 - 132988974 on Build GRCh38 | Loss |
|
| esv3893774 | Chr.3:132705936 - 132933476 on Build GRCh38 | Loss |
|
| esv3597830 | Chr.3:132728764 - 132733663 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs73003824] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map