Assay Details
Target Gene Details
Entrez Gene ID: | 134466 |
Gene Name: | zinc finger protein 300 pseudogene 1 |
Gene Aliases: |
- |
Location: |
Chr.5:150930436-150946584 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF300P1 | NR_026867.1 | |||
| AK091436.1 | ||||
| AK096536.1 | ||||
| AK097462.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv526742 | Chr.5:150932060 - 150952038 on Build GRCh38 | Gain |
|

Genomic Map