Assay Details
Target Gene Details
Entrez Gene ID: | 5558 |
Gene Name: | primase (DNA) subunit 2 |
Gene Aliases: |
PRIM2A, p58 |
Location: |
Chr.6:57314805-57646852 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRIM2 | NM_000947.4 | NP_000938.2 | ||
| NM_001282487.1 | NP_001269416.1 | |||
| NM_001282488.1 | NP_001269417.1 | |||
| XM_017011004.1 | XP_016866493.1 | |||
| XM_017011005.1 | XP_016866494.1 | |||
| XM_017011006.1 | XP_016866495.1 | |||
| XM_017011007.1 | XP_016866496.1 | |||
| AK223287.1 | ||||
| AK291915.1 | ||||
| BC017833.1 | AAH17833.1 | |||
| BC064931.1 | AAH64931.1 | |||
| BI547222.1 | ||||
| X74331.1 | CAA52378.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759432 | Chr.6:57319743 - 60594857 on Build GRCh38 | Gain |
|
| esv3609117 | Chr.6:57231347 - 57341646 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map