Assay Details
Target Gene Details
Entrez Gene ID: | 7291 |
Gene Name: | twist family bHLH transcription factor 1 |
Gene Aliases: |
ACS3, BPES2, BPES3, CRS, CRS1, CSO, SCS, TWIST, bHLHa38 |
Location: |
Chr.7:19113047-19117672 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TWIST1 | XM_011515496.1 | XP_011513798.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3612462 | Chr.7:19111202 - 19116559 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map