Assay Details
Target Gene Details
Entrez Gene ID: | 142685 |
Gene Name: | ankyrin repeat and SOCS box containing 15 |
Gene Aliases: |
- |
Location: |
Chr.7:123567003-123637900 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ASB15 | NM_001290258.1 | NP_001277187.1 | ||
| NM_080928.4 | NP_563616.3 | |||
| XM_005250146.4 | XP_005250203.1 | |||
| XM_011515816.2 | XP_011514118.1 | |||
| XM_011515817.2 | XP_011514119.1 | |||
| XM_011515818.2 | XP_011514120.1 | |||
| XM_011515819.2 | XP_011514121.1 | |||
| XM_011515820.2 | XP_011514122.1 | |||
| XM_011515821.2 | XP_011514123.1 | |||
| XM_017011756.1 | XP_016867245.1 | |||
| XM_017011757.1 | XP_016867246.1 | |||
| XM_017011758.1 | XP_016867247.1 | |||
| AF428257.1 | AAP97297.1 | |||
| AK125360.1 | BAC86146.1 | |||
| BC100874.2 | AAI00875.1 | |||
| BC100875.2 | AAI00876.1 | |||
| BC100876.1 | ||||
| BC100877.2 | AAI00878.1 | |||
| CR749595.1 | CAH18393.1 |
Target Gene Details
Entrez Gene ID: | 102724555 |
Gene Name: | uncharacterized LOC102724555 |
Gene Aliases: |
- |
Location: |
Chr.7:123614161-123624957 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC102724555 | NR_111922.1 | |||
| DB024487.1 | ||||
| DB050194.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3891198 | Chr.7:121885430 - 123886905 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map