Assay Details
Target Gene Details
Entrez Gene ID: | 222008 |
Gene Name: | V-set and transmembrane domain containing 2A |
Gene Aliases: |
VSTM2 |
Location: |
Chr.7:54542064-54571080 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| VSTM2A | NM_001301009.1 | NP_001287938.1 | ||
| NM_182546.3 | NP_872352.2 | |||
| NR_133927.1 | ||||
| XM_006715665.3 | XP_006715728.1 | |||
| XM_006715666.3 | XP_006715729.1 | |||
| XM_017011841.1 | 5 | 1836 | XP_016867330.1 | |
| BI667944.1 | ||||
| BI752069.1 | ||||
| BI754879.1 | ||||
| BI758069.1 | ||||
| BX451729.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2734425 | Chr.7:53868335 - 55917254 on Build GRCh38 | Deletion |
|
| nsv1024361 | Chr.7:53993970 - 54776794 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map